Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180990.3 | NP_851321.2 | 2 | 213 | ||
GenBank mRNA | AB223030.1 | - | 2 | 127 | ||
AF512521.1 | - | 2 | 167 | |||
AK122638.1 | - | 2 | 213 | |||
BC110596.1 | - | 2 | 167 | |||
BC110597.1 | - | 2 | 167 | 40023955 | ||
DB119720.1 | - | 2 | 213 |