Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 4 | 804 | ||
NM_002629.3 | NP_002620.1 | 4 | 874 | |||
GenBank mRNA | AK223096.1 | - | 4 | 720 | ||
AK292216.1 | - | 4 | 775 | |||
AK293023.1 | - | 4 | 719 | |||
AK295991.1 | - | 5 | 610 | |||
AK296619.1 | - | 4 | 663 | |||
AK297491.1 | - | 5 | 620 | |||
AK312351.1 | - | 4 | 703 | |||
AK315910.1 | - | 4 | 807 | |||
AY007118.1 | - | 1 | 711 | |||
BC000455.1 | - | 1 | 737 | 2821567 | ||
BC010038.1 | - | 1 | 729 | 3604026 | ||
BC011678.2 | - | 1 | 693 | 4299008 | ||
BC053356.1 | - | 1 | 710 | 5787678 | ||
BC062302.1 | - | 1 | 709 | 6584466 | ||
BC066959.1 | - | 4 | 742 | 5262935 | ||
BC073742.1 | - | 1 | 708 | 6297173 | ||
BC157873.1 | - | 4 | 726 | 8862699 | ||
CR541659.1 | - | 4 | 691 | |||
CR541744.1 | - | 4 | 691 | |||
EU794644.1 | - | 4 | 703 | |||
J04173.1 | - | 4 | 722 |
Gene Symbol: PGAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029891.2 | NP_001025062.1 | 1 | 691 |