Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017564.9 | NP_060034.9 | 11, 12 | 1443 | ||
XM_011538537.2 | XP_011536839.1 | 11, 12 | 1484 | |||
XM_011538538.2 | XP_011536840.1 | 11, 12 | 1484 | |||
XM_017019585.1 | XP_016875074.1 | 11, 12 | 1484 | |||
GenBank mRNA | AB052958.1 | - | 11, 12 | 1439 | ||
AJ295695.1 | - | 11, 12 | 1443 | |||
AK160380.1 | - | 10, 11 | 1158 | |||
AY311388.1 | - | 11, 12 | 1461 |