Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 5 | 753 | |||
GenBank mRNA | AK057241.1 | - | 5 | 672 | ||
BC144672.1 | - | 3 | 450 | |||
BC150302.1 | - | 5 | 727 | 8860399 | ||
DQ309038.1 | - | 5 | 642 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 5 | 753 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 5 | 753 | |||
GenBank mRNA | AK292534.1 | - | 5 | 639 | ||
BC110417.1 | - | 5 | 724 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 5 | 753 | |||
GenBank mRNA | AK310240.1 | - | 5 | 727 | ||
BC063309.1 | - | 5 | 727 | |||
DQ309037.1 | - | 5 | 727 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 5 | 753 | |||
GenBank mRNA | AK302813.1 | - | 5 | 747 | ||
BC066774.1 | - | 5 | 727 | |||
BC099913.1 | - | 4 | 1580 | 4838463 | ||
BC125193.1 | - | 4 | 545 | 40122746 | ||
BC125194.1 | - | 4 | 545 | 40122749 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 5 | 727 |