Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172105.1 | NP_001165576.1 | 13 | 2526 | ||
NM_017577.4 | NP_060047.3 | 18 | 2799 | |||
XM_005247546.2 | XP_005247603.1 | 17 | 2715 | |||
XM_005247547.1 | XP_005247604.1 | 17 | 2584 | |||
XM_011512930.1 | XP_011511232.1 | 18 | 2794 | |||
XM_011512931.1 | XP_011511233.1 | 13 | 2583 | |||
XM_017006646.1 | XP_016862135.1 | 16 | 2536 | |||
XM_017006647.1 | XP_016862136.1 | 19 | 2877 | |||
XM_017006648.1 | XP_016862137.1 | 14 | 2606 | |||
XM_017006649.1 | XP_016862138.1 | 12 | 2357 | |||
GenBank mRNA | AL133661.1 | - | 9 | 3015 | ||
AY358725.1 | - | 13 | 2479 | |||
BC028972.2 | - | 10 | 1803 | |||
BC035040.1 | - | 18 | 2805 | 4829130 | ||
BX648633.1 | - | 2 | 2836 | |||
CV575069.1 | - | 1 | 120 |