Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251107
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
7
995
NM_001184901.1
NP_001171830.1
7
942
NM_001184902.1
NP_001171831.1
10
1358
NM_001184903.1
NP_001171832.1
8
1092
NM_014959.3
NP_055774.2
8
1053
NR_033678.1
6
912
NR_033679.1
6
954
NR_033680.1
6
912
XM_006723091.3
XP_006723154.1
9
1206
XM_006723092.3
XP_006723155.1
11
1744
XM_006723093.3
XP_006723156.1
9
1527
XM_006723094.3
XP_006723157.1
9
1186
XM_006723095.3
XP_006723158.1
9
1173
XM_006723096.3
XP_006723159.1
10
1595
XM_006723097.3
XP_006723160.1
8
1378
XM_006723098.3
XP_006723161.1
8
1165
XM_006723099.3
XP_006723162.1
7
1051
XM_006723101.2
XP_006723164.1
8
1068
XM_006723102.3
XP_006723165.1
7
1008
XM_006723104.3
XP_006723167.1
8
1065
XM_006723106.3
XP_006723169.1
9
1538
XM_006723107.3
XP_006723170.1
9
1189
XM_006723109.3
XP_006723172.1
8
1065
XM_011526641.2
XP_011524943.1
12
2051
XM_011526642.2
XP_011524944.1
11
1946
XM_011526643.2
XP_011524945.1
10
1633
XM_011526644.2
XP_011524946.1
8
1218
XM_011526646.2
XP_011524948.1
9
1692
XM_011526647.2
XP_011524949.1
8
1581
XM_011526648.2
XP_011524950.1
9
1698
XM_011526650.2
XP_011524952.1
9
1189
XM_011526653.2
XP_011524955.1
9
1728
XM_017026482.1
XP_016881971.1
9
1531
XM_017026483.1
XP_016881972.1
9
1484
XM_017026484.1
XP_016881973.1
9
1492
XM_017026485.1
XP_016881974.1
9
1189
XM_017026486.1
XP_016881975.1
7
1048
XM_017026487.1
XP_016881976.1
6
1417
XM_017026488.1
XP_016881977.1
10
1633
XM_017026489.1
XP_016881978.1
9
1218
XM_017026490.1
XP_016881979.1
9
1189
XM_017026491.1
XP_016881980.1
9
1189
XM_017026492.1
XP_016881981.1
9
1500
XM_017026493.1
XP_016881982.1
8
1392
XM_017026494.1
XP_016881983.1
9
2006
XM_017026495.1
XP_016881984.1
8
1148
XM_017026496.1
XP_016881985.1
8
1068
XM_017026497.1
XP_016881986.1
7
1008
XM_017026498.1
XP_016881987.1
7
1005
XM_017026499.1
XP_016881988.1
6
2778
GenBank mRNA
AB023172.1
-
6
947
AF322184.1
-
6
947
AF331519.1
-
4
634
AF405558.1
-
4
634
AF511652.1
-
6
947
AK297045.1
-
8
1078
AK297069.1
-
7
932
AK302327.1
-
6
882
AY026322.1
-
4
634
BX641091.1
-
3
865
BX648568.1
-
8
3099
EU118120.1
-
7
995
EU118121.1
-
6
954
EU118122.1
-
4
795
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0