Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3947 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 3013 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3814 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3501 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 4076 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4340 | |||
NM_003879.5 | NP_003870.4 | 10 | 3919 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4814 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3783 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3862 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4749 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6766 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4622 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3960 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3585 |
Gene Symbol: ACOT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB014607.1 | - | 17 | 4223 |
Gene Symbol: ERVK-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ304053.1 | - | 1 | 111 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336887.1 | - | 1 | 303 | ||
AF338196.1 | - | 1 | 321 | |||
AF338199.1 | - | 1 | 276 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 53455 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1581 |
Gene Symbol: RRM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | H70418.1 | - | 1 | 59 |
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 462 |
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832327.2 | - | 12 | 3094 |