Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 2 | 163 | ||
XM_005256769.1 | XP_005256826.1 | 2 | 163 | |||
XM_005256770.1 | XP_005256827.1 | 2 | 163 | |||
XM_011523993.1 | XP_011522295.1 | 2 | 163 | |||
XM_011523994.1 | XP_011522296.1 | 2 | 163 | |||
XM_011523995.1 | XP_011522297.1 | 2 | 163 | |||
GenBank mRNA | AK315818.1 | - | 2 | 150 | ||
BC073889.1 | - | 2 | 132 | 5222492 | ||
DA426549.1 | - | 2 | 163 |
Gene Symbol: LGALS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042685.1 | NP_001036150.1 | 2 | 158 | ||
GenBank mRNA | AK290263.1 | - | 2 | 159 | ||
BC105943.1 | - | 2 | 135 | 40031793 | ||
DA668702.1 | - | 2 | 158 |