Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198950.1 | NP_001185879.1 | 26, 27 | 3748 | ||
NM_015011.1 | NP_055826.1 | 26, 27 | 3186 | |||
XM_011521062.1 | XP_011519364.1 | 26, 27 | 3294 | |||
GenBank mRNA | AB020672.2 | - | 26, 27 | 3186 | ||
AB290159.1 | - | 26, 27 | 3749 | |||
AK095691.1 | - | 21, 22 | 2446 | |||
AK302933.1 | - | 14, 15 | 1889 | |||
BC146791.1 | - | 26, 27 | 3186 | 9007256 |