Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 12 | 1874 | ||
NM_010589.6 | NP_034719.2 | 12 | 1874 | |||
GenBank mRNA | AK038268.1 | - | 11 | 1685 | ||
AK088365.1 | - | 12 | 1723 | |||
AK142178.1 | - | 12 | 1712 | |||
AK164520.1 | - | 12 | 1714 | |||
BC105577.1 | - | 11 | 1677 | 3968386 | ||
BC131646.1 | - | 11 | 1685 | 40110146 | ||
BC131647.1 | - | 11 | 1685 | 40110148 | ||
L32955.1 | - | 11 | 1673 | |||
L33768.1 | - | 9 | 2315 | |||
L40172.1 | - | 12 | 1735 |