Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251300
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
11
800
NM_001001568.1
NP_001001568.1
9
651
NM_001001569.1
NP_001001569.1
8
596
NM_001001570.1
NP_001001570.1
11
821
NM_001001571.1
NP_001001571.1
10
729
NM_001001572.1
NP_001001572.1
8
607
NM_001001573.1
NP_001001573.1
9
787
NM_001001574.1
NP_001001574.1
10
722
NM_001001575.1
NP_001001575.1
7
552
NM_001001576.1
NP_001001576.1
7
511
NM_001001577.1
NP_001001577.1
8
630
NM_001001578.1
NP_001001578.1
9
674
NM_001001579.1
NP_001001579.1
9
667
NM_001001580.1
NP_001001580.1
6
467
NM_001001581.1
NP_001001581.1
10
831
NM_001001582.1
NP_001001582.1
11
909
NM_001001583.1
NP_001001583.1
11
902
NM_001001584.2
NP_001001584.1
10
730
NM_001001585.1
NP_001001585.1
10
854
NM_001315533.1
NP_001302462.1
12
876
NM_002606.2
NP_002597.1
12
980
XM_011529598.2
XP_011527900.1
12
902
XM_011529600.2
XP_011527902.1
12
953
XM_017028366.1
XP_016883855.1
10
718
GenBank mRNA
AF048837.1
-
12
977
AF067223.1
-
12
929
AF067224.1
-
11
748
AF067225.1
-
9
590
AF067226.1
-
8
535
AK001165.1
-
9
582
AK001307.1
-
11
771
AK127770.1
-
2
2331
AK294112.1
-
9
617
AK308753.1
-
6
456
AK314679.1
-
12
953
AY196299.1
-
11
794
AY196300.1
-
10
702
AY196301.1
-
8
580
AY196302.1
-
9
760
AY196303.1
-
10
695
AY196304.1
-
7
525
AY196305.1
-
7
484
AY196306.1
-
8
603
AY196307.1
-
9
647
AY196308.1
-
9
640
AY196309.1
-
6
440
AY196310.1
-
10
804
AY196311.1
-
11
882
AY196312.1
-
11
875
AY196313.1
-
10
702
AY196314.1
-
10
827
AY242121.1
-
10
666
AY701187.1
-
12
873
BC009047.1
-
11
774
3874635
BT007016.1
-
11
737
KU178246.1
-
11
737
KU178247.1
-
12
917
KU178248.1
-
8
533
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0