Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 1701 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 1718 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 1713 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 1839 | |||
NM_001548.4 | NP_001539.3 | 2 | 1604 | |||
GenBank mRNA | AK092813.1 | - | 2 | 2632 | ||
AK308688.1 | - | 3 | 1605 | |||
BC007091.1 | - | 2 | 1529 | 4250452 | ||
X03557.1 | - | 2 | 1501 |