Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sssca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020491.5 | NP_065237.3 | 1, 2 | 58 | ||
GenBank mRNA | AB012865.1 | - | 1, 2 | 26 | ||
AK005265.1 | - | 1, 2 | 30 | |||
AK012592.1 | - | 1, 2 | 36 | |||
AK012704.1 | - | 1, 2 | 36 | |||
BC021593.1 | - | 1, 2 | 28 | 5102521 | ||
BY081504.1 | - | 1, 2 | 116 |