Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177296.4 | NP_796270.2 | 4 | 624 | ||
GenBank mRNA | AK045143.1 | - | 4 | 652 | ||
AK049446.1 | - | 4 | 838 | |||
AK145723.1 | - | 4 | 641 | |||
BC064646.1 | - | 4 | 624 | 6313452 | ||
BC075678.1 | - | 4 | 837 | 30619729 |