Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ranbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177721.4 | NP_808389.2 | 1 | 580 | ||
GenBank mRNA | AK044143.1 | - | 1 | 565 | ||
BC056759.1 | - | 1 | 559 | 6518485 | ||
BC139395.1 | - | 1 | 580 | 8862417 | ||
BC139396.1 | - | 1 | 580 | 8862418 |