Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001719.1 | NP_001001719.1 | 2, 3 | 209 | ||
GenBank mRNA | AY621075.1 | - | 2, 3 | 209 |