Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001938.3 | NP_001001938.1 | 3 | 3804 | ||
NM_001142413.1 | NP_001135885.1 | 3 | 3747 | |||
GenBank mRNA | AK127854.1 | - | 3 | 3739 | ||
AK130650.1 | - | 1 | 1277 | |||
AL831887.1 | - | 1 | 1173 |