Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 3 | 401 | ||
GenBank mRNA | AF548565.1 | - | 3 | 344 | ||
AK135814.1 | - | 593 |