Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 25 | 2740 | ||
GenBank mRNA | AF548565.1 | - | 25 | 2683 | ||
AK075830.1 | - | 3 | 192 | |||
BC046963.1 | - | 25 | 2678 | |||
BC100666.1 | - | 20 | 2046 |