Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004339.2 | NP_001004339.2 | 11 | 2277 | ||
NM_001307931.1 | NP_001294860.1 | 10 | 1525 | |||
XM_011541480.2 | XP_011539782.1 | 10 | 2007 | |||
XM_011541481.2 | XP_011539783.1 | 10 | 1875 | |||
XM_011541484.1 | XP_011539786.1 | 11 | 2052 | |||
GenBank mRNA | AY271826.1 | - | 10 | 1516 |