Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 14, 15 | 4863 | ||
XM_011512928.2 | XP_011511230.1 | 14, 15 | 4954 | |||
GenBank mRNA | AB033094.1 | - | 9, 10 | 2833 | ||
AK022542.1 | - | 2, 3 | 668 | |||
AK026003.1 | - | 73 | ||||
AY134858.1 | - | 9, 10 | 4036 | |||
BC032804.1 | - | 1, 2 | 579 | |||
BX648758.1 | - | 10, 11 | 4181 | |||
DQ063584.1 | - | 11, 12 | 4327 | |||
DQ063585.1 | - | 11, 12 | 4327 |