Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 19 | 2891 | ||
NM_001171944.1 | NP_001165415.1 | 17 | 2641 | |||
NM_025154.5 | NP_079430.3 | 18 | 2754 | |||
GenBank mRNA | AB018353.1 | - | 20 | 2989 | ||
AK022816.1 | - | 18 | 2746 | |||
AK025737.1 | - | 16 | 2416 | |||
AK074320.1 | - | 6 | 1338 | |||
AK128170.1 | - | 3 | 2376 | |||
BC142707.1 | - | 19 | 2901 | 40148817 | ||
BC152418.1 | - | 19 | 2989 | 8862528 | ||
BX538211.1 | - | 5 | 740 | |||
BX640866.1 | - | 10 | 2980 | |||
BX648893.1 | - | 10 | 2980 | |||
BX649069.1 | - | 17 | 2646 |