Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009621.2 | NP_001009621.1 | 4 | 1403 | ||
GenBank mRNA | BC088299.1 | - | 4 | 1403 | 7323100 | |
FQ216253.1 | - | 4 | 1189 |