Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009631.1 | NP_001009631.1 | 80 | |||
GenBank mRNA | BC088319.1 | - | 80 | 7325542 |