Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009643.2 | NP_001009643.2 | 2 | 208 | ||
XM_006228995.1 | XP_006229057.1 | 2 | 208 | |||
GenBank mRNA | BC088252.1 | - | 2 | 208 | 7372248 |