Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009669.1 | NP_001009669.1 | 6 | 690 | ||
GenBank mRNA | BC088757.1 | - | 6 | 690 | 7309521 |