Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011924.1 | NP_001011924.1 | 14 | 2445 | ||
XM_006254909.2 | XP_006254971.1 | 16 | 2887 | |||
XM_006254910.2 | XP_006254972.1 | 16 | 2884 | |||
XM_006254913.2 | XP_006254975.1 | 15 | 2718 | |||
GenBank mRNA | BC081932.1 | - | 14 | 2445 | 7114194 |