Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011964.1 | NP_001011964.1 | 9, 10 | 1647 | ||
XM_006233949.2 | XP_006234011.1 | 10, 11 | 1833 | |||
XM_006233950.2 | XP_006234012.1 | 9, 10 | 1385 | |||
XM_008761720.1 | XP_008759942.1 | 8, 9 | 1324 | |||
XM_008761721.1 | XP_008759943.1 | 8, 9 | 1347 | |||
XM_008761722.1 | XP_008759944.1 | 7, 8 | 1257 | |||
XM_008761723.1 | XP_008759945.1 | 7, 8 | 935 | |||
GenBank mRNA | BC079073.1 | - | 9, 10 | 1647 | 7122012 |