Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011984.1 | NP_001011984.1 | 10 | 2054 | ||
XM_006240464.2 | XP_006240526.1 | 8 | 2699 | |||
GenBank mRNA | BC085882.1 | - | 10 | 2054 | 7191983 |
Gene Symbol: LOC100909439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764916.1 | XP_008763138.1 | 5 | 1132 |