Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193509.1 | NP_001180438.1 | 2 | 155 | ||
NM_017735.4 | NP_060205.3 | 2 | 402 | |||
XM_005264416.1 | XP_005264473.1 | 2 | 389 | |||
XM_011532958.1 | XP_011531260.1 | 2 | 389 | |||
GenBank mRNA | AK027570.1 | - | 2 | 311 | ||
AK299196.1 | - | 2 | 144 | |||
BC063791.1 | - | 2 | 360 | 6500951 | ||
BI546117.1 | - | 2 | 414 |