Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012346.1 | NP_001012346.1 | 1 | 382 | ||
XM_008771429.1 | XP_008769651.1 | 3 | 830 | |||
XM_008771430.1 | XP_008769652.1 | 3 | 830 | |||
XM_008771431.1 | XP_008769653.1 | 2 | 677 | |||
XM_008771432.1 | XP_008769654.1 | 2 | 669 | |||
XM_008771433.1 | XP_008769655.1 | 2 | 542 | |||
GenBank mRNA | AJ810421.1 | - | 1 | 382 |