Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013052.1 | NP_001013070.1 | 1 | 247 | ||
XM_006253021.2 | XP_006253083.1 | 2 | 2444 |