Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013078.1 | NP_001013096.1 | 2 | 210 | ||
XM_006234734.2 | XP_006234796.1 | 2 | 245 | |||
XM_006234735.2 | XP_006234797.1 | 2 | 245 | |||
XM_008762090.1 | XP_008760312.1 | 3 | 188 | |||
GenBank mRNA | BC082080.1 | - | 2 | 210 | 7133293 |