Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017443.1 | NP_001017443.1 | 2, 3 | 285 | ||
XM_006253141.2 | XP_006253203.1 | 2, 3 | 201 | |||
XM_006253143.2 | XP_006253205.1 | 3, 4 | 1282 | |||
XM_008771257.1 | XP_008769479.1 | 2, 3 | 201 | |||
GenBank mRNA | BC092575.1 | - | 2, 3 | 285 | 7114719 |