Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017995.2 | NP_001017995.1 | 13 | 3367 | ||
XM_017009351.1 | XP_016864840.1 | 14 | 3451 | |||
GenBank mRNA | AB037716.1 | - | 2 | 2114 |