Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024238.1 | NP_001019409.1 | 10 | 1044 | ||
XM_006250425.2 | XP_006250487.1 | 9 | 1180 | |||
XM_006250426.2 | XP_006250488.1 | 9 | 1180 | |||
XM_006250427.2 | XP_006250489.1 | 5 | 560 | |||
GenBank mRNA | AY383705.1 | - | 10 | 1044 |