Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024743.1 | NP_001019914.1 | 4, 5 | 681 | ||
XM_006251538.1 | XP_006251600.1 | 4, 5 | 633 | |||
GenBank mRNA | BC097369.1 | - | 4, 5 | 681 | 7443949 |