Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021766.1 | NP_068534.1 | 3 | 1337 | ||
GenBank mRNA | AF163321.1 | - | 3 | 1293 | ||
BC062073.1 | - | 3 | 1297 | 5599449 | ||
U63315.1 | - | 3 | 1337 |