Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012301.2 | NP_001012301.1 | 2 | 2260 | ||
GenBank mRNA | AB448735.1 | - | 2 | 2251 | ||
AK122641.1 | - | 2 | 1404 | |||
AY875937.1 | - | 2 | 1671 | |||
BC129995.1 | - | 2 | 1728 | 40127932 | ||
BC129996.1 | - | 2 | 1728 | 40127934 |