Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012302.2 | NP_001012302.2 | 8 | 739 | ||
XM_011520048.1 | XP_011518350.1 | 8 | 739 | |||
XM_011520051.2 | XP_011518353.1 | 8 | 739 | |||
XM_011520052.2 | XP_011518354.1 | 8 | 739 | |||
XM_011520053.2 | XP_011518355.1 | 8 | 739 | |||
XM_017017644.1 | XP_016873133.1 | 8 | 739 | |||
XM_017017645.1 | XP_016873134.1 | 8 | 739 | |||
XM_017017646.1 | XP_016873135.1 | 8 | 739 | |||
XM_017017647.1 | XP_016873136.1 | 8 | 739 | |||
GenBank mRNA | AK026923.1 | - | 8 | 686 | ||
AK074088.1 | - | 5 | 2152 | |||
AK303642.1 | - | 7 | 781 | |||
AK316479.1 | - | 7 | 807 | |||
BC128557.1 | - | 8 | 707 | 40122929 | ||
BC144415.1 | - | 7 | 765 |