Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013180.1 | NP_001013198.1 | 1 | 14 | ||
GenBank mRNA | AF436841.1 | - | 1 | 14 |