Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013407.2 | NP_001013425.2 | 4 | 1551 | ||
GenBank mRNA | BC101344.1 | - | 4 | 1537 | 40025152 |
Gene Symbol: PRAMEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010889.2 | NP_001010889.1 | 4 | 1537 | ||
GenBank mRNA | BC101342.2 | - | 4 | 1537 | 40025149 |