Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300816.2 | NP_001287745.1 | 5 | 1717 | ||
NM_001300817.2 | NP_001287746.1 | 5 | 1699 | |||
NM_001300818.2 | NP_001287747.1 | 4 | 1675 | |||
NM_001300819.2 | NP_001287748.1 | 3 | 1609 | |||
NM_001300822.2 | NP_001287751.1 | 5 | 1745 | |||
NM_001308127.1 | NP_001295056.1 | 5 | 1747 | |||
NM_001308128.1 | NP_001295057.1 | 3 | 1489 | |||
NM_001308129.1 | NP_001295058.1 | 2 | 1447 | |||
NM_001308130.1 | NP_001295059.1 | 4 | 1679 | |||
NM_017665.3 | NP_060135.1 | 4 | 1651 | |||
NR_125337.2 | 4 | 1655 | ||||
NR_131766.1 | 5 | 1800 | ||||
NR_131767.1 | 4 | 1683 | ||||
XM_011543502.1 | XP_011541804.1 | 4 | 1704 | |||
XM_011543503.2 | XP_011541805.1 | 5 | 1792 | |||
GenBank mRNA | AK000101.1 | - | 4 | 1595 | ||
AK122970.1 | - | 5 | 1644 |