Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014979.2 | NP_001014979.2 | 2, 3 | 731 | ||
NM_001195620.1 | NP_001182549.1 | 2, 3 | 731 | |||
XM_011545976.1 | XP_011544278.1 | 2, 3 | 764 | |||
XM_011545978.1 | XP_011544280.1 | 2, 3 | 764 | |||
XM_017023852.1 | XP_016879341.1 | 2, 3 | 764 | |||
XM_017023854.1 | XP_016879343.1 | 2, 3 | 764 | |||
XM_017023855.1 | XP_016879344.1 | 2, 3 | 764 | |||
XM_017023856.1 | XP_016879345.1 | 2, 3 | 285 | |||
XM_017023857.1 | XP_016879346.1 | 1, 2 | 941 | |||
XM_017023858.1 | XP_016879347.1 | 2, 3 | 764 | |||
XM_017023859.1 | XP_016879348.1 | 2, 3 | 764 | |||
XM_017023861.1 | XP_016879350.1 | 2, 3 | 764 | |||
XM_017023862.1 | XP_016879351.1 | 1, 2 | 941 | |||
GenBank mRNA | AK307279.1 | - | 1, 2 | 889 | ||
AL122109.2 | - | 1, 2 | 1000 | |||
BC042548.1 | - | 1, 2 | 785 | |||
BC073881.1 | - | 2, 3 | 565 | |||
BC092491.1 | - | 1, 2 | 1023 | 4837696 | ||
BC128128.1 | - | 2, 3 | 235 | |||
BC128129.1 | - | 2, 3 | 235 | 40112368 | ||
BI829598.1 | - | 2, 3 | 727 |