Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017924.3 | NP_001017924.1 | 2 | 293 | ||
NM_001284223.1 | NP_001271152.1 | 2 | 293 | |||
GenBank mRNA | BC140765.1 | - | 2 | 293 | 9021633 | |
BC144475.1 | - | 2 | 293 | 9053002 | ||
BX648524.1 | - | 2 | 233 |