Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024927.2 | NP_001020098.1 | 5 | 544 | ||
NM_001081641.1 | NP_001075110.1 | 4 | 496 | |||
GenBank mRNA | AK039594.1 | - | 5 | 433 | ||
AK147444.1 | - | 5 | 501 | |||
AK147630.1 | - | 4 | 329 | |||
BC112411.1 | - | 4 | 919 | 40061037 |