Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032538.1 | NP_115927.1 | 11, 12 | 1522 | ||
XM_017011366.1 | XP_016866855.1 | 10, 11 | 1312 | |||
GenBank mRNA | AB058758.1 | - | 9, 10 | 1091 | ||
AB218664.1 | - | 12, 13 | 1326 | |||
AK131217.1 | - | 10, 11 | 1252 |