Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017694.3 | NP_060164.3 | 8 | 2892 | ||
XM_005246654.2 | XP_005246711.1 | 7 | 2770 | |||
XM_005246656.3 | XP_005246713.1 | 8 | 2795 | |||
XM_011511368.1 | XP_011509670.1 | 9 | 2958 | |||
XM_017004392.1 | XP_016859881.1 | 9 | 2917 | |||
GenBank mRNA | AK000167.1 | - | 6 | 1670 | ||
AK021420.1 | - | 4 | 360 | |||
AK098085.1 | - | 6 | 2302 | |||
AK127176.1 | - | 8 | 2892 | |||
AK225240.1 | - | 6 | 1670 | |||
AK302647.1 | - | 6 | 1170 | |||
AK302753.1 | - | 7 | 1127 | |||
BC050537.2 | - | 6 | 1831 | 6204874 |