Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF536205.1 | - | 3686 | |||
AK026371.1 | - | 1474 | ||||
BC006556.2 | - | 2740 | ||||
BC048345.1 | - | 1648 | ||||
BC064981.1 | - | 3540 | 6585236 |