Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 5 | 620 | ||
GenBank mRNA | AA168368.1 | - | 2 | 76 | ||
AB009661.1 | - | 5 | 620 | |||
AK007843.1 | - | 5 | 619 | |||
AK131898.1 | - | 5 | 619 | |||
AK160484.1 | - | 5 | 619 | |||
BC139220.1 | - | 5 | 705 | 8862242 | ||
BC139223.1 | - | 5 | 705 | 8862245 |